Learn More
Enzo Life Sciences COP9 signalosome Csn7 subunit (human) polyclonal antibody (25 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW83000025
Encompass_Preferred
Immunogen: Recombinant full length human COP9 signalosome (Csn7 subunit). Species reactivity: Human. Specificity: Recognizes the Csn7 subunit of the COP9 signalosome. Applications: IP, WB. Formulation: Liquid. In PBS containing 10mM sodium azide.
Catalog No. 50-201-1618