Learn More
Enzo Life Sciences COP9 signalosome Csn2 subunit polyclonal antibody (25 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW82300025
Encompass_Preferred
Immunogen: Recombinant N-terminal COP9 signalosome (Csn2 subunit). Species reactivity: Human. Specificity: Recognizes the Csn2 subunit of the COP9 signalosome. Applications: IHC, IP, WB. Formulation: Liquid. In PBS containing 10mM sodium azide.
Catalog No. 50-201-1599