Learn More
Enzo Life Sciences COP9 signalosome Csn3 subunit polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW82350100
Encompass_Preferred
Immunogen: Recombinant human COP9 signalosome (Csn3 subunit) (aa 115-345). Species reactivity: Human. Specificity: Recognizes the Csn3 subunit of the COP9 signalosome. Applications: IHC, WB. Formulation: Liquid. In PBS containing 10mM sodium azide.
Catalog No. 50-201-1602