Learn More
Aviva Systems Biology Corp COPA Antibody - middle region (ARP51894_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51894_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against COPA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human COPA. Alternate names: FLJ26320, HEP-COPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60640