Learn More
Copan Diagnostics Calibrated Disposable Inoculating Loops and Needles in Zip Lock Pack
GSA_VASDP
Catalog No. 1490629 Shop All Copan Diagnostics Inc. Products
Change view
Click to view available options
Product Type:
Inoculating Loop, Flexible
Inoculating Loop, Rigid
Inoculating Needle
Color:
Dark Blue
Dark Green
Light Blue
Light Green
Violet
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Color
14-906-29 Inoculating Loop, Flexible Light Green
14-906-30 Inoculating Loop, Flexible Light Blue
14-906-28 Inoculating Loop, Rigid Dark Blue
14-906-27 Inoculating Loop, Rigid Dark Green
14-906-31 Inoculating Needle Violet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 14-906-29 Supplier Copan Diagnostics Supplier No. COPS1
Add to Cart
Add to Cart
This item is not returnable. View return policy
Calibrated 1 and 10μL plastic disposable inoculating loops and needles
Calibrated 1 and 10μL plastic disposable inoculating loops and needles
Zip-lock closure reseals the pack between use to avoid contamination. Re-sealable bag allows the user to open and close the loop pack to remove individual loops.
  • Disposable, sterile plastic loops and needles manufactured from soft-flexible or hard-rigid plastic
  • Plastic injection molding technology ensures product consistency and eliminates variations in loop thickness and internal diameter
  • Ultrasmooth surfaces allow easy streaking without cutting or gouging agar surface
  • Calibrated using the Evans Blue Dye Method recommended by the American Society for Microbiology
  • Color coded for easy product identification
  • Packed in tamper-proof bags to guarantee sterility
  • Certificate of calibration included in each case
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Disposable Single-use
Volume 1 μL
Sterility Sterile
Material Plastic
Color Light Green
Product Type Inoculating Loop, Flexible
Includes Evans blue dye certificate of calibration
Packaging Type Tamperproof Ziplock resealable bag
Calibrated Yes