Learn More
Copan Diagnostics Capture Cap for ESwab™ Tube
SDP
Catalog No. 23600530 Shop All Copan Diagnostics Inc. Products
Change view
Click to view available options
Color:
Red
Catalog No. Color
23-600-530 Red
1 options
Catalog No. 23-600-530 Supplier Copan Diagnostics Supplier No. PFT1R913R
Add to Cart
Add to Cart
Securely attach the swab applicator stick to the cap. The Copan Diagnostics Capture Cap for ESwab™ Tube allows the operator to conveniently remove the swab and perform various microbiology analyses using the tube cap as a handle to hold and manipulate the swab.
Specifications
Color Red
Product Type Component Capture Cap