Learn More
Copan Diagnostics ESwab™ MRSA Collection System
SDP
Catalog No. 23600904 Shop All Copan Diagnostics Inc. Products
Change view
Click to view available options
Includes:
Pink Polypropylene screw-cap tube with internal conical shape filled with 1mL of Liquid Amies Medium, 1 pink regular size flock swab plus 1 white regular size flocked swab
Pink Polypropylene screw-cap tube with internal conical shape filled with 1mL of Liquid Amies Medium, 2 pink regular size flock swab plus 1 white regular size flocked swab
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
23-600-904 Pink Polypropylene screw-cap tube with internal conical shape filled with 1mL of Liquid Amies Medium, 2 pink regular size flock swab plus 1 white regular size flocked swab
23-600-903 Pink Polypropylene screw-cap tube with internal conical shape filled with 1mL of Liquid Amies Medium, 1 pink regular size flock swab plus 1 white regular size flocked swab
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-600-904 Supplier Copan Diagnostics Supplier No. 493C03
Add to Cart
Add to Cart
This item is not returnable. View return policy
Sample collection pack for various microbiology analyses
  • Sterile
  • Single use
  • Pink Polypropylene screw-cap
  • Applicator stick securely attaches to the cap after collection
  • Tube with internal conical shape filled with 1mL of Liquid Amies Medium
  • Sampling sites: Nose, throat, perineum
  • 50 units per shelf pack
  • 10 x 50 kits per box
Specifications
Sterility Sterile
Includes Pink Polypropylene screw-cap tube with internal conical shape filled with 1mL of Liquid Amies Medium, 2 pink regular size flock swab plus 1 white regular size flocked swab
For Use With (Application) For various microbiology analysis
Product Type MRSA Collection System