Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copan Diagnostics Nylon Flocked Dry Swabs in Peel Pouches
GSA_VASDP

Catalog No. 23600955 Shop All Copan Diagnostics Inc. Products
Change view
Click to view available options
Length (English) Shaft:
1.97 in.
3.15 in.
3.94 in.
Length (Metric) Shaft:
50 mm
80 mm
100 mm
Tip Type:
Contoured Adult
Contoured Pediatric
Flexible Minitip
Minitip flocked
Regular flocked
Ultra minitip/pediatric
Ultra-thin/minitip
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Shaft Length (Metric) Shaft Tip Type
23-600-955 3.94 in. 100 mm Ultra minitip/pediatric
23-600-967 1.97 in. 50 mm Contoured Pediatric
23-600-968 3.15 in. 80 mm Contoured Pediatric
23-600-966 3.15 in. 80 mm Contoured Adult
23-600-958 3.15 in. 80 mm Ultra-thin/minitip
23-600-950 3.15 in. 80 mm Minitip flocked
23-600-951 3.15 in. 80 mm Regular flocked
23-600-956 3.94 in. 100 mm Minitip flocked
23-600-957 3.94 in. 100 mm Regular flocked
23-600-952 3.94 in. 100 mm Flexible Minitip
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 23-600-955 Supplier Copan Diagnostics Supplier No. 516CS01
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Permits complete, rapid elution by keeping the entire sample close to the surface. Copan Diagnostics Nylon Flocked Dry Swabs in Peel Pouches are anatomically designed swabs to optimize analyte collection efficiency with improved patient comfort.

Sterile, individually wrapped swabs in a plastic applicator.
  • Ergonomic and Anatomic Design: Improves patient comfort and specimen collection
  • Rapid Automatic Elution: Instant and spontaneous release of sample into liquid media
  • Measurable and Consistent Uptake and Transfer: Quantitative measurement and improved test sensitivity
  • Patient Friendly: Anatomical design and a soft texture for rapidly and efficiently dislodging cells
  • Compatible with Multiple Applications: Rapid antigen testing, EIA, molecular-based assays, DFA, cytology testing, bacteriology and virology culture
  • Safe and Convenient: Molded breakpoint

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Applicator Material Plastic
Tip Type Ultra minitip/pediatric
Sterility Sterile
Packaging Individually-wrapped in peel pouch
Length (Metric) Shaft 100 mm
Length (English) Shaft 3.94 in.
Product Type Specimen Collection Swab
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.