Learn More
Copan Diagnostics Nylon-Flocked Dry Swabs in Tubes
GSA_VASDP
Catalog No. 23600963 Shop All Copan Diagnostics Inc. Products
Change view
Click to view available options
Tip Type:
Flexible Minitip Flocked
Minitip Flocked
Pediatric
Regular Flocked
Ultra-thin/Minitip
Breakpoint:
80 mm
100 mm
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Type Breakpoint
23-600-963 Minitip Flocked 100 mm
23-600-962 Pediatric 100 mm
23-600-964 Regular Flocked 100 mm
23-600-959 Minitip Flocked 80 mm
23-600-960 Regular Flocked 80 mm
23-600-965 Ultra-thin/Minitip 80 mm
23-600-961 Flexible Minitip Flocked 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 23-600-963 Supplier Copan Diagnostics Supplier No. 518C
Add to Cart
Add to Cart
This item is not returnable. View return policy
Permit easy, rapid elution
  • Plastic applicator
  • Sterile swabs in a dry tube
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Tip Type Minitip Flocked
Applicator Material Plastic
Sterility Sterile
Breakpoint 100 mm
Product Type Specimen Collection Swab