Learn More
Copan Diagnostics Nylon-Flocked Dry Swabs in Tubes
SDP
Catalog No. 23600965 Shop All Copan Diagnostics Inc. Products
Change view
Click to view available options
Tip Type:
Flexible Minitip Flocked
Minitip Flocked
Pediatric
Regular Flocked
Ultra-thin/Minitip
Breakpoint:
80 mm
100 mm
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Type Breakpoint
23-600-965 Ultra-thin/Minitip 80 mm
23-600-963 Minitip Flocked 100 mm
23-600-962 Pediatric 100 mm
23-600-964 Regular Flocked 100 mm
23-600-959 Minitip Flocked 80 mm
23-600-960 Regular Flocked 80 mm
23-600-961 Flexible Minitip Flocked 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 23-600-965 Supplier Copan Diagnostics Supplier No. 560C
Add to Cart
Add to Cart
Permit easy, rapid elution
  • Plastic applicator
  • Sterile swabs in a dry tube
Specifications
Breakpoint 80 mm
Product Type Specimen Collection Swab
Sterility Sterile
Applicator Material Plastic
Tip Type Ultra-thin/Minitip