Learn More
Copan Diagnostics Plain Dry Swabs in Labeled Tubes
GSA_VASDP
Catalog No. 1490619 Shop All Copan Diagnostics Inc. Products
Change view
Click to view available options
Applicator Material:
Aluminum
Plastic
Twisted Wire
Color:
Blue
Orange
Red
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Applicator Material Color
14-906-19 Plastic Red
14-906-22 Aluminum Orange
14-906-23 Aluminum Yellow
14-906-25 Twisted Wire Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 14-906-19 Supplier Copan Diagnostics Supplier No. 159C
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Sterile plain dry swabs in labeled tubes
  • Choice of applicator and tip styles
  • Color coded
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Applicator Material Plastic
Tip Material Polyester
Sterility Sterile
Packaging Labeled tubes
Color Red
Product Type Specimen Collection Swab