Learn More
Copan Diagnostics Plain Dry Swabs in Labeled Tubes
SDP
Catalog No. 1490622 Shop All Copan Diagnostics Inc. Products
Change view
Click to view available options
Applicator Material:
Aluminum
Plastic
Twisted Wire
Color:
Blue
Orange
Red
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Applicator Material Color
14-906-22 Aluminum Orange
14-906-19 Plastic Red
14-906-23 Aluminum Yellow
14-906-25 Twisted Wire Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 14-906-22 Supplier Copan Diagnostics Supplier No. 160C
Add to Cart
Add to Cart
This item is not returnable. View return policy
Sterile plain dry swabs in labeled tubes
  • Choice of applicator and tip styles
  • Color coded
Specifications
Tip Material Rayon
Tip Type Minitip flocked
Applicator Material Aluminum
Sterility Sterile
Packaging Labeled tubes
Color Orange
Product Type Specimen Collection Swab