Learn More
Copan Diagnostics Plain Dry Swabs in Peel Pouches
GSA_VASDP
Catalog No. 1490640 Shop All Copan Diagnostics Inc. Products
Change view
Click to view available options
Applicator Material:
Aluminum
Plastic
Tip Material:
Polyester
Rayon
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Applicator Material Tip Material
14-906-40 Aluminum Rayon
14-906-50 Aluminum Polyester
14-906-41 Plastic Polyester
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 14-906-40 Supplier Copan Diagnostics Supplier No. 170KS01
Add to Cart
Add to Cart
This item is not returnable. View return policy
Sterile plain dry swabs with regular tips in space-efficient peel pouches. 10 boxes x 100 swabs
Single Regular Size Traditional Dry Polyester Swab with Plastic Applicator packaged with Peel Pouch - Individually Packaged, Sterile
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Applicator Material Aluminum
Sterility Sterile
Tip Material Rayon
Packaging Individually-wrapped in peel pouch
No. per Pack 100 / Box
No. per Case 10 Boxes x 100 Swabs / Case
Product Type Specimen Collection Swab