Learn More
COPAN FecalSwab™
GSA_VA
Catalog No. R723487 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 Kits/Pk.
Catalog No. Quantity
R723487 50 Kits/Pk.
1 options
This item is not returnable. View return policy
Catalog No. R723487 Supplier Thermo Scientific Supplier No. R723487
Add to Cart
Add to Cart
This item is not returnable. View return policy
Simplify fecal specimen collection and preservation for enteric diagnostics with the COPAN FecalSwab™ System, designed for automated specimen processing.
Optimize specimen collection and preservation for enteric diagnostics with the COPAN FecalSwab™ System. An advanced liquid-based collection and preservation system for fecal specimens, COPAN FecalSwab™, is easy-to-use, allowing for sample collection from either rectal swabbing or direct from feces. Its versatility makes possible direct plating or aliquot transfer to selective enrichment broth. COPAN FecalSwab™ converts solid or semi-solid fecal specimens into a sample suitable for automatic specimen processors in space-saving, instrument-ready tubes. It is also suitable for PCR.
Simplify and standardize collection and transport of feces and rectal swabs with compact, neat and easy-to-transport COPAN FecalSwab™s.
  • Easy-to-use - suitable for both rectal swabbing or directly from feces. Small tube is instrument-ready and easy- to- transport.
  • Optimized - flocked swab aids adequate specimen collection and elution of the sample into medium. When opening the tube in the lab, swab is captured by the cap, for easier access to the specimen.
  • Versatile - sample can be planted and streaked directly on culture plates or an aliquot transferred to selective enrichment broth. It is also suitable for PCR.
  • Efficient - suitable for automated specimen processing
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Media Type Cary-Blair
Closure Color Orange
Description FecalSwab™ Regular Flocked Collection Kit
For Use With (Application) Rectal and Fecal Specimens
Quantity 50 Kits/Pk.
Product clearance and availability restrictions may apply in some countries.
IVD – For in vitro diagnostic use.
FecalSwab™ is a trademark of COPAN DIAGNOSTICS, INC.