Learn More
TARGETMOL CHEMICALS INC COPANLISIB 25MG
SDP
Catalog No. 502576532
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Copanlisib (BAY 80-6946) is a phosphoinositide 3-kinase (PI3K) inhibitor with potential antineoplastic activity. Copanlisib inhibits the activation of the PI3K signaling pathway which may result in inhibition of tumor cell growth and survival in susceptible tumor cell populations. Activation of the PI3K signaling pathway is frequently associated with tumorigenesis and dysregulated PI3K signaling may contribute to tumor resistance to a variety of antineoplastic agents. purity: 99%
Catalog No. 50-257-6532 Supplier TARGETMOL CHEMICALS INC Supplier No. T632225MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Copanlisib (BAY 80-6946) is a phosphoinositide 3-kinase (PI3K) inhibitor with potential antineoplastic activity. Copanlisib inhibits the activation of the PI3K signaling pathway which may result in inhibition of tumor cell growth and survival in susceptible tumor cell populations. Activation of the PI3K signaling pathway is frequently associated with tumorigenesis and dysregulated PI3K signaling may contribute to tumor resistance to a variety of antineoplastic agents. purity: 99%