Learn More
Medchemexpress LLC HY-15346A 5mg Medchemexpress, Copanlisib (dihydrochloride) CAS:1402152-13-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15346A5MG
Encompass_Preferred
Medchemexpress, HY-15346A 5mg Copanlisib (dihydrochloride) CAS:1402152-13-9 Copanlisib dihydrochloride (BAY 80-6946 dihydrochloride) is a potent, selective and ATP-competitive pan-class I PI3K inhibitor, with IC50s of 0.5 nM, 0.7 nM, 3.7 nM and 6.4 nM for PI3Kα, PI3Kδ, PI3Kβ and PI3Kγ, respectively. Copanlisib dihydrochloride has more than 2,000-fold selectivity against other lipid and protein kinases, except for mTOR. Copanlisib dihydrochloride has superior antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0614