Learn More
Apexbio Technology LLC Copanlisib dihydrochloride 1402152-13-9 10mg
SDP
Catalog No. 5000096014
Encompass_Preferred
Copanlisib dihydrochloride (CAS 1402152-13-9) is a small-molecule inhibitor that selectively targets phosphoinositide 3-kinase (PI3K) isoforms and By inhibiting these PI3K subunits copanlisib disrupts PI3K/AKT signaling pathways that regulate cellular proliferation and survival leading to reduced tumor cell growth and induction of apoptosis This compound is utilized in biomedical research to investigate PI3K-mediated oncogenic processes particularly in studies related to relapsed or refractory follicular lymphoma and other hematologic malignancies dependent on aberrant PI3K activity
Catalog No. 50-000-96014 Supplier Apexbio Technology LLC Supplier No. A992610
Copanlisib dihydrochloride (CAS 1402152-13-9) is a small-molecule inhibitor that selectively targets phosphoinositide 3-kinase (PI3K) isoforms and By inhibiting these PI3K subunits copanlisib disrupts PI3K/AKT signaling pathways that regulate cellular proliferation and survival leading to reduced tumor cell growth and induction of apoptosis This compound is utilized in biomedical research to investigate PI3K-mediated oncogenic processes particularly in studies related to relapsed or refractory follicular lymphoma and other hematologic malignancies dependent on aberrant PI3K activity