Learn More
Medchemexpress LLC Copanlisib dihydrochloride | 1402152-13-9 | 99.6% | 553.44 | C23H30Cl2N8O4 | 10 MG
SDP
Catalog No. 5000286794
Encompass_Preferred
Copanlisib dihydrochloride is a potent, selective pan-class I PI3K inhibitor supplied as a solid research reagent for preclinical and in-vitro studies. It is provided in a small-scale 10 mg pack and includes characterization data such as molecular weight, formula, and reported purity to support experimental planning.
  • Potent, selective pan-class I PI3K inhibition.
  • High purity suitable for research (99.6%).
  • Supplied as a solid powder for convenient handling and storage.
  • Recommended storage: 4°C sealed; in solvent: -80°C (6 months) or -20°C (1 month).
  • Documented physicochemical identifiers for traceability.
Catalog No. 50-002-86794 Supplier Medchemexpress LLC Supplier No. HY15346A10MG
Copanlisib dihydrochloride is a potent, selective pan-class I PI3K inhibitor supplied as a solid research reagent for preclinical and in-vitro studies. It is provided in a small-scale 10 mg pack and includes characterization data such as molecular weight, formula, and reported purity to support experimental planning.
  • Potent, selective pan-class I PI3K inhibition.
  • High purity suitable for research (99.6%).
  • Supplied as a solid powder for convenient handling and storage.
  • Recommended storage: 4°C sealed; in solvent: -80°C (6 months) or -20°C (1 month).
  • Documented physicochemical identifiers for traceability.