Learn More
CPC Scientific H-Ala-Ser-Asp-Arg-Ser-Asn-Ala-Thr-Gln-Leu-Asp-Gly-Pro-Ala-Gly-Ala-Leu-Leu-Leu-Arg-Leu-Val-Gln-Leu-Ala-Gly-Ala-Pro-Glu-Pro-Phe-Glu-Pro-Ala-Gln-Pro-Asp-Ala-Tyr-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific COPE001A
Encompass
This peptide derived from the vasopressin-neurophysin 2-copeptin precursor, a prognostic marker in a variety of diseases, such as sepsis, community-acquired pneumonia, chronic obstructive pulmonary failure, heart failure and myocardial infarction.ONE-LETTER SEQUENCE: ASDRSNATQLDGPAGALLLRLVQLAGAPEPFEPAQPDAYMOLECULAR FORMULA: C177H279N49O58MOLECULAR WEIGHT:4021.46STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [78362-34-2], RESEARCH AREA: CardiovascularREFERENCES: J.Struck et al., Peptides, 26, 2500 (2005); N.G.Morgenthaler et al., Clin. Chem., 52, 112 (2006); S.Q.Khan et al., Circulation, 115, 2103 (2007); G.Szinnai et al., J. Clin. Endocrinol. Metab., 92, 3973 (2007); M.Katan et al., Crit. Care, 12, 117 (2008); R.Seligman et al., Crit. Care, 12, R11 (2008)
Catalog No. 50-210-9135