Learn More
Biomatik Corporation Copeptin (Cpp) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU31041
Encompass_Preferred
Copeptin (Cpp) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Copeptin (CPP). The Copeptin (Cpp) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Asp128~Leu144. Applications: WB, IHC, ICC, IP. Target Synonyms: Copeptin (CPP). Accession Number: P01185. Gene ID: 551. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5012