Learn More
Aviva Systems Biology Corp COPG2 Antibody - N-terminal region (ARP54845_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54845_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against COPG2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human COPG2. Alternate names: 2-COP, FLJ11781ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61340