Learn More
Med Vet International Coplin Glass Jar, J0814X
Catalog No. 5000582213
Encompass_Preferred
Clear thick flint glass with internal ridges for separating slides from each other.
Catalog No. 50-005-82213 Supplier Med Vet International Supplier No. JOR814X
Clear thick flint glass with internal ridges for separating slides from each other.