Learn More
Alkali Scientific Coplin Glass Staining Jar with Glass Lid, 5 Slide Capacity, for 25×75 mm Slides, 20/Case
SDP
Catalog No. 5000662829
Encompass_Preferred
This Coplin style glass staining jar is designed for routine staining and processing of standard 25.0 × 75.0 mm (1" × 3") microscope slides in histology and laboratory applications. The jar holds up to 5 slides in an upright orientation, allowing uniform exposure to staining reagents during laboratory procedures. Constructed from high quality glass, it provides excellent chemical resistance and clear visibility of contents during use. The included glass lid helps minimize evaporation and contamination while maintaining reagent stability. This jar is ideal for histology, pathology, and general slide staining workflows requiring consistent and reliable reagent contact.
Catalog No. 50-006-62829 Supplier Alkali Scientific Supplier No. SJ005CS
This Coplin style glass staining jar is designed for routine staining and processing of standard 25.0 × 75.0 mm (1" × 3") microscope slides in histology and laboratory applications. The jar holds up to 5 slides in an upright orientation, allowing uniform exposure to staining reagents during laboratory procedures. Constructed from high quality glass, it provides excellent chemical resistance and clear visibility of contents during use. The included glass lid helps minimize evaporation and contamination while maintaining reagent stability. This jar is ideal for histology, pathology, and general slide staining workflows requiring consistent and reliable reagent contact.