Learn More
Epredia™ Coplin Staining Jar
Catalog No. 194 Shop All Epredia Products
Change view
Click to view available options
Slide Count:
10
Catalog No. Slide Count
19-4 10
1 options
Catalog No. 19-4 Supplier Epredia™ Supplier No. 194
Add to Cart
Add to Cart
Hold 10 slides back-to-back with the Epredia™ Plastic Coplin Staining Jar.
Features:
  • Holds 10 slides back-to-back
  • 1.6 in. (40mm) diameter circular tapered top
  • High-density polyethylene construction with internal grooves
Includes: Domed screw-on cap
Specifications
Description Plastic Coplin Staining Jar
Material Plastic
Type Coplin Staining Jar
Includes Domed screw-on cap
Slide Count 10
Not intended to be microwavable