Learn More
Andwin Scientific COPLIN STAINING JAR
SDP
Catalog No. NC9727395
Encompass
The Glass Coplin Staining Jar can store 5 single slides measuring 1 x 3" vertically, or 10 slides back-to-back. The slides remain within the jar so that these can be capped by the included screw cap during staining procedures. This product is sold in a case of 4/
Catalog No. NC9727395 Supplier Andwin Scientific Supplier No. 70317
The Glass Coplin Staining Jar can store 5 single slides measuring 1 x 3" vertically, or 10 slides back-to-back. The slides remain within the jar so that these can be capped by the included screw cap during staining procedures. This product is sold in a case of 4/