Learn More
Electron Microscopy Sciences Coplin Staining Jar, Screw Cap
SDP
Catalog No. NC0202223
Encompass_Preferred
5-Slide Unit. Holds 5 single 1"x3" slides vertically, or 10 slides back-to-back. Slides extend above the opening, allowing for the manipulation of them without the use of forceps. A screw cap has a metal foil liner that withstands stains and organic solvents and it prevents evaporation.
Catalog No. NC0202223 Supplier Electron Microscopy Sciences Supplier No. 70315
5-Slide Unit. Holds 5 single 1"x3" slides vertically, or 10 slides back-to-back. Slides extend above the opening, allowing for the manipulation of them without the use of forceps. A screw cap has a metal foil liner that withstands stains and organic solvents and it prevents evaporation.