Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Copolyester, Carboy
GREENER_CHOICE

Catalog No. 19830448 Shop All Justrite Safety Group Products
Change view
Click to view available options
Capacity (Metric):
2.5 L
5 L
10 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
19-830-448 10 L
19-830-446 2.5 L
19-830-447 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-830-448 Supplier Justrite™ Supplier No. 12948
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Use With: For storing and dispensing solvents, buffers, reagents and powders. Especially when visual inspection needed.; Closure Material: Polypropylene; Color: Clear

  • Constructed from clear, BPA-free, USP Class VI, FDA grade resin
  • Rectangular ergonomic design
  • Interchangeable caps and adapters provide top connection flexibility
  • Material, volume, and metric graduation marks imprinted on ca

Specifications

Accuracy ±5%
Capacity (English) 2.6 gal.
Capacity (Metric) 10 L
Closure Material Polypropylene
Closure Size 83 mm
Color Clear
Dimensions (L x W x H) 273 x 163 x 495 mm
For Use With (Application) For storing and dispensing solvents, buffers, reagents and powders. Especially when visual inspection needed.
Height (English) 19.5 in.
Length (English) 10.75 in.
Material Copolyester
Product Type Carboy
Shape Rectangular
Width (English) 6.4 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.