Learn More
Research Products International Corp 7 ml Copolymer Plastic Vial, Unlined White Poly Screw Cap, Bulk Pack, 1000 per Case
SDP
Catalog No. 50213002
Encompass
High clarity plastic vial for liquid scintillation counting. Supplied with an unlined white polyethylene cap that seals with 1/4 turn. Each bulk pack contains 1,000 vials and caps packaged in separate clear plastic bags. These may not be compatible with certain scintillation counter models that require cap and vial to be flush.
Catalog No. 50-213-002 Supplier Research Products International Corp Supplier No. 125509
High clarity plastic vial for liquid scintillation counting. Supplied with an unlined white polyethylene cap that seals with 1/4 turn. Each bulk pack contains 1,000 vials and caps packaged in separate clear plastic bags. These may not be compatible with certain scintillation counter models that require cap and vial to be flush.