Learn More
Medchemexpress LLC Copovidone | 25086-89-9 | 50.0% | 45,000-70,000 g/mol | (C6H9NO.C4H6O2)n | 25 G
SDP
Catalog No. 5000285173
Encompass_Preferred
Copovidone (50% in ethanol) is a vinylpyrrolidone-vinyl acetate copolymer supplied as a 50% (w/v) solution in ethanol. It functions as a pharmaceutical excipient used as a film former, binder, and solubilizing agent in formulation development and laboratory testing. Supplied in small laboratory quantities for research and excipient evaluation.
  • 50% solution in ethanol for ready-to-use formulations.
  • Acts as a film former and binder in coating and tablet applications.
  • Improves solubility and drug loading for poorly soluble compounds.
  • Suitable for formulation development and analytical testing.
  • Available in small laboratory sizes appropriate for research use.
Catalog No. 50-002-85173 Supplier Medchemexpress LLC Supplier No. HYW392209A25G
Copovidone (50% in ethanol) is a vinylpyrrolidone-vinyl acetate copolymer supplied as a 50% (w/v) solution in ethanol. It functions as a pharmaceutical excipient used as a film former, binder, and solubilizing agent in formulation development and laboratory testing. Supplied in small laboratory quantities for research and excipient evaluation.
  • 50% solution in ethanol for ready-to-use formulations.
  • Acts as a film former and binder in coating and tablet applications.
  • Improves solubility and drug loading for poorly soluble compounds.
  • Suitable for formulation development and analytical testing.
  • Available in small laboratory sizes appropriate for research use.