Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Standard, 0.1 ppm Cu in 12.5% (v/v) Ethanol, Ricca Chemical
SDP

Catalog No. R2279010500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R2279010500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R2279010500 Supplier Ricca Chemical Company Supplier No. R2279010500A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 64-17-5 , 7697-37-2 , 7758-99-8

Specifications

Packaging Natural Poly Bottle
CAS Min % 89.0, 9.27, 0.01
CAS Max % 92.0, 9.65, 0.01
Physical Form Liquid
Color Blue to Colorless
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Analytical Standard
Quantity 500 mL
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.1ppm in 12.5% v/v Ethanol
Chemical Name or Material Copper Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.