Learn More
Medchemexpress LLC Copper(4+), [[N,N',N'',N'''-(29H,31H-phthalocyanine-C,C,C,C-tetrayl-κN29,κN30,κN31,κN32)tetrakis[methylenethio[(dimethylamino)methylidyne]]]tetrakis[N-methylmethanaminiumato]](2-)]-, chloride (1:4) | 33864-99-2 | 50 MG
SDP
Catalog No. 5000410566
Encompass_Preferred
Alcian Blue 8GX is a commonly used phthalocyanine dye that binds to glycoproteins and glycosaminoglycans. It is utilized across a wide range of biological staining applications.
  • Binds to glycoproteins and glycosaminoglycans.
  • Used for staining proteins in brain tumors.
  • Applies to staining DNA in cells and tissues.
Catalog No. 50-004-10566 Supplier Medchemexpress LLC Supplier No. HYD000150MG
Alcian Blue 8GX is a commonly used phthalocyanine dye that binds to glycoproteins and glycosaminoglycans. It is utilized across a wide range of biological staining applications.
  • Binds to glycoproteins and glycosaminoglycans.
  • Used for staining proteins in brain tumors.
  • Applies to staining DNA in cells and tissues.