Learn More
Electron Microscopy Sciences Omniprobe FIB Lift-Out Grid 5-Post, Copper
SDP
Catalog No. 5033765
Encompass
Custom copper 5-post lift-out grids specially designed for in-situ lift-out. These grids include multiple indexed mounting locations, all with vertical bars attachment surfaces. Now with lower profile sides for easier access to outermost posts 3 mm diameter.
Material: Copper
Catalog No. 50-337-65 Supplier Electron Microscopy Sciences Supplier No. 7596404
Custom copper 5-post lift-out grids specially designed for in-situ lift-out. These grids include multiple indexed mounting locations, all with vertical bars attachment surfaces. Now with lower profile sides for easier access to outermost posts 3 mm diameter.
Material: Copper