Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper aluminum oxide, 99.5% (metals basis)

Catalog No. AA3941930
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3941930 250 g
AA3941918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3941930 Supplier Thermo Scientific Chemicals Supplier No. 03941930
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 12042-92-1 | Al2CuO4 | 181.51 g/mol

Applications for copper aluminum oxide nanocrystalsare included in microbatteries, in micro-electronics, circuits and in nanowire, and in nanofluids.It is also a promising material for thermoelectric devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12042-92-1
Molecular Formula Al2CuO4
Molecular Weight (g/mol) 181.51
MDL Number MFCD00210610
InChI Key IPJRADJOSNLGBF-UHFFFAOYSA-N
PubChem CID 12947132
IUPAC Name copper;oxido(oxo)alumane
SMILES [Cu++].[O-][Al]=O.[O-][Al]=O

Specifications

Physical Form Powder
Quantity 250 g
Solubility Information Insoluble in water.
Formula Weight 181.5
Percent Purity 99.5%
Assay (metals basis)
Chemical Name or Material Copper aluminum oxide
EINECSNumber 234-934-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.