Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Beryllium foil, alloy C172, 0.13mm (0.005in) thick

Catalog No. AA41911GU
Change view
Click to view available options
Quantity:
150 x 200 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41911GU 150 x 200 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA41911GU Supplier Thermo Scientific Chemicals Supplier No. 041911GU
Add to Cart

missing translation for 'validMainframeMsg'

 

Uses include springs of all kinds where high strength and corrosion resistance are important, resistance welding electrodes, dies and high strength, corrosion resistant tubing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Copper Beryllium foil, Alloy C172
Name Note 0.13mm (0.005 in.) thick, Temper: half hard
Avg. Mol. Wt. or Mol. Wt. Range Cu:Be; 98.1:1.9 wt%
Quantity 150 x 200 mm
Odor Odorless
Weight ≈32.6g/150x200mm
Form Foil
MDL Number MFCD01320550
Solubility Information Insoluble in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.