Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Buffer, Ricca Chemical
SDP

Catalog No. R2265000020 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
20 L
Packaging:
Cubitainer
Hard Pack
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R2265000020 20 L Hard Pack
R22650001A 1 L Natural Poly Bottle
R226500020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R2265000020 Supplier Ricca Chemical Company Supplier No. R226500020E1
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Copper Buffer
CAS 12125-02-9, 1336-21-6, 7732-18-5
Color Colorless
Concentration 1X
For Use With (Application) Water Analysis
pH 10.3
Physical Form Liquid
Quantity 20 L
Solution Type Buffer
Odor characteristic ammonia odor
Packaging Hard Pack
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.