Learn More
Strem, An Ascensus Company CAS# 12018-10-9. 100g. Copper chromite, barium promoted (62-64% Cr2CuO4, 22-24% CuO, 6%BaO, 0-4% Graphite, 1% CrO3, 1% Cr2O3). MFCD00148884
SDP
Catalog No. 5090111294
Encompass
CAS# 12018-10-9. 100g. Copper chromite, barium promoted (62-64% Cr2CuO4, 22-24% CuO, 6%BaO, 0-4% Graphite, 1% CrO3, 1% Cr2O3). MFCD00148884. Molecular Weight: 79.55/231.53. Molecular Formula: CuO/Cr2CuO4. Color/form: black pellets Strem# 29-0410. http://www.strem.com/catalog/v/29-0410/
Catalog No. 50-901-11294 Supplier Strem, An Ascensus Company Supplier No. 290410100G
CAS# 12018-10-9. 100g. Copper chromite, barium promoted (62-64% Cr2CuO4, 22-24% CuO, 6%BaO, 0-4% Graphite, 1% CrO3, 1% Cr2O3). MFCD00148884. Molecular Weight: 79.55/231.53. Molecular Formula: CuO/Cr2CuO4. Color/form: black pellets Strem# 29-0410. http://www.strem.com/catalog/v/29-0410/