Learn More
Bioworld Copper-Coated Microplates, 5 Plate(s)
Catalog No. 50197545
Encompass
Copper coated microlates are ideal for analyzing polyhistidine-tagged fusion proteins by ELISA-based methods. Proteins that contain a succession of several histidine residues at the amino or carboxyl terminus have a strong binding affinity for metals such as copper. These copper-coated plates are available in clear that can be used with colorimetric methods.Specifications:For quantitatingpreviously undetectable histidine-tagged proteinsCopper chelate provides greater binding capacity than nickel Detection limit is 0.1 pg of polyhistidine tagged fusion protein per well Pre-blocked for the ultimate convenience
Catalog No. 50-197-545 Supplier Bioworld Supplier No. 201400141
Copper coated microlates are ideal for analyzing polyhistidine-tagged fusion proteins by ELISA-based methods. Proteins that contain a succession of several histidine residues at the amino or carboxyl terminus have a strong binding affinity for metals such as copper. These copper-coated plates are available in clear that can be used with colorimetric methods.Specifications:For quantitatingpreviously undetectable histidine-tagged proteinsCopper chelate provides greater binding capacity than nickel Detection limit is 0.1 pg of polyhistidine tagged fusion protein per well Pre-blocked for the ultimate convenience