Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Copper, dichloro[(2Z)-2-[(4-oxo-4H-1-benzopyran-3-yl)methylene] hydrazinecarbothioamide-κN2,κS]- | 902779-59-3 | 95.0% | 381.73 g·mol⁻¹ | C11H9Cl2CuN3O2S | 10 MG
SDP

Catalog No. 5000286443
Encompass_Preferred

FPA-124 is a cell-permeable copper complex research reagent that selectively inhibits Akt (protein kinase B) with an IC50 of 0.1 μM. It interacts with both the pleckstrin homology and kinase domains of Akt and has been reported to induce apoptosis in cell-based studies. Intended for research use only.

  • Selective Akt inhibition (IC50 = 0.1 μM).
  • Cell-permeable copper complex.
  • Reported to induce apoptosis in cell-based assays.
  • Purity: 95.0%.
  • Molecular weight: 381.73 g·mol⁻¹.
  • Available in small pack sizes suitable for research (mg scale).

Catalog No. 50-002-86443 Supplier Medchemexpress LLC Supplier No. HY1536910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FPA-124 is a cell-permeable copper complex research reagent that selectively inhibits Akt (protein kinase B) with an IC50 of 0.1 μM. It interacts with both the pleckstrin homology and kinase domains of Akt and has been reported to induce apoptosis in cell-based studies. Intended for research use only.

  • Selective Akt inhibition (IC50 = 0.1 μM).
  • Cell-permeable copper complex.
  • Reported to induce apoptosis in cell-based assays.
  • Purity: 95.0%.
  • Molecular weight: 381.73 g·mol⁻¹.
  • Available in small pack sizes suitable for research (mg scale).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.