Learn More
Analytical Sales & Services Copper Electrode. Features 2 holes.
Catalog No. 5000805133
Encompass
Copper Electrode\nFeatures 2 holes.  For use as cathode in undivided flow cell, either cathode or anode in divided flow cell.\n\n• Thickness: 3mm\n• Height: 50mm\n• Width: 50mm\n\nQty/Pkg: Each
Catalog No. 50-008-05133 Supplier Analytical Sales & Services Supplier No. F1088149
Copper Electrode\nFeatures 2 holes.  For use as cathode in undivided flow cell, either cathode or anode in divided flow cell.\n\n• Thickness: 3mm\n• Height: 50mm\n• Width: 50mm\n\nQty/Pkg: Each