Learn More
Medchemexpress LLC Copper Fluor-4 | 2365532-64-3 | 98.4% | 749.05 | 1 MG
SDP
Catalog No. 5000359407
Encompass_Preferred
Copper Fluor-4 (CF4) is a Cu+-specific fluorescent probe based on a rhodol dye scaffold. It exhibits high selectivity for copper (Kd value of 2.9x10-13 M), especially over zinc, iron, and various cellular alkali and alkaline earth metals. The probe maintains stability within a physiologically relevant pH range of 6 to 8, with excitation at 415 nm and emission at 660 nm.
  • Specific fluorescent probe for Cu+ ions
  • Developed from a rhodol dye scaffold
  • Offers high selectivity for copper, including over zinc and iron
  • Maintains stability across a pH range of 6 to 8
  • Excitation at 415 nm
  • Emission at 660 nm
  • Can localize copper in the soma region of PC-12-derived neurons
  • Suitable for assessing labile copper distributions in living organisms like zebrafish embryos
  • Monitors labile Cu pools through reversible Cu binding in worms
Catalog No. 50-003-59407 Supplier Medchemexpress LLC Supplier No. HY1500861MG
Copper Fluor-4 (CF4) is a Cu+-specific fluorescent probe based on a rhodol dye scaffold. It exhibits high selectivity for copper (Kd value of 2.9x10-13 M), especially over zinc, iron, and various cellular alkali and alkaline earth metals. The probe maintains stability within a physiologically relevant pH range of 6 to 8, with excitation at 415 nm and emission at 660 nm.
  • Specific fluorescent probe for Cu+ ions
  • Developed from a rhodol dye scaffold
  • Offers high selectivity for copper, including over zinc and iron
  • Maintains stability across a pH range of 6 to 8
  • Excitation at 415 nm
  • Emission at 660 nm
  • Can localize copper in the soma region of PC-12-derived neurons
  • Suitable for assessing labile copper distributions in living organisms like zebrafish embryos
  • Monitors labile Cu pools through reversible Cu binding in worms