Learn More
Strem, An Ascensus Company CAS# 7440-50-8. 200 x 200mm. Copper foil (99%). MFCD00010965
SDP
Catalog No. 5090114944
Encompass
CAS# 7440-50-8. 200 x 200mm. Copper foil (99%). MFCD00010965. Molecular Weight: 63.54. Molecular Formula: Cu. Color/form: 0.127 mm thick x 200 mm wide Strem# 93-2992. http://www.strem.com/catalog/v/93-2992/
Catalog No. 50-901-14944 Supplier Strem, An Ascensus Company Supplier No. 932992200MM
CAS# 7440-50-8. 200 x 200mm. Copper foil (99%). MFCD00010965. Molecular Weight: 63.54. Molecular Formula: Cu. Color/form: 0.127 mm thick x 200 mm wide Strem# 93-2992. http://www.strem.com/catalog/v/93-2992/