Learn More
Glass Expansion Copper Foil for Ceramic Outer Tube PerkinElmer Optima 8000,8300 DV Series, 31-808-2990 (Pack of 12)
SDP
Catalog No. 502703136
Encompass_Preferred
Copper Foil for Ceramic Outer Tube (Pack of 12). Compatible with Glass Expansion’s Ceramic Outer Tube for D-Torch (501907170, 31-808-2990) for PerkinElmer Optima 8000,8300 DV Series.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3136 Supplier Glass Expansion Supplier No. 318083356
Copper Foil for Ceramic Outer Tube (Pack of 12). Compatible with Glass Expansion’s Ceramic Outer Tube for D-Torch (501907170, 31-808-2990) for PerkinElmer Optima 8000,8300 DV Series.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001