Learn More
Glass Expansion Copper foil for torch body (Pack of 12)
SDP
Catalog No. 502703155
Encompass_Preferred
Copper Foil for Torch Body (N077-5297, N0775297, 310-20, 8003-0363). Package of 12 copper foils. Compatible with quartz torches for PerkinElmer Optima 2000/4000/5000/7000 DV ICP-OES.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3155 Supplier Glass Expansion Supplier No. 318082645
Copper Foil for Torch Body (N077-5297, N0775297, 310-20, 8003-0363). Package of 12 copper foils. Compatible with quartz torches for PerkinElmer Optima 2000/4000/5000/7000 DV ICP-OES.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001