Learn More
Strem, An Ascensus Company CAS# 598-54-9. 2g. Copper(I) acetate, 97%. MFCD00058908
SDP
Catalog No. NC0325248
Encompass
CAS# 598-54-9. 2g. Copper(I) acetate, 97%. MFCD00058908. Molecular Weight: 122.59. Molecular Formula: CuOOCCH3. Color/form: beige to light green pwdr. Strem# 93-2989. http://www.strem.com/catalog/v/93-2989/
Catalog No. NC0325248 Supplier Strem, An Ascensus Company Supplier No. 9329892G
CAS# 598-54-9. 2g. Copper(I) acetate, 97%. MFCD00058908. Molecular Weight: 122.59. Molecular Formula: CuOOCCH3. Color/form: beige to light green pwdr. Strem# 93-2989. http://www.strem.com/catalog/v/93-2989/