Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(I) bromide, 98%, extra pure, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
Packaging:
Glass bottle
7787-70-4
BrCu
143.45
MFCD00010969
NKNDPYCGAZPOFS-UHFFFAOYSA-M
copper i bromide,copper bromide cubr,copper 1+ bromide,copper monobromide,cuprousbromide,hsdb 270,cubr,copper 1 bromide
24593
bromocopper
[Cu]Br
7787-70-4
BrCu
143.45
MFCD00010969
NKNDPYCGAZPOFS-UHFFFAOYSA-M
copper i bromide,copper bromide cubr,copper 1+ bromide,copper monobromide,cuprousbromide,hsdb 270,cubr,copper 1 bromide
24593
bromocopper
[Cu]Br
Assay Percent Range 97.5% min. (Complexometry)
Linear Formula CuBr
Fieser 01,165; 02,90; 03,67; 04,108; 05,163; 06,143; 07,79; 08,116; 11,140; 12,286; 14,99; 16,96
Merck Index 15, 2649
Arsenic (As) 50ppm max.
Solubility Information Solubility in water: soluble in cold water. Other solubilities: decomposes in hot water,sol. in hcl,hbr,nh4oh (formation of complexes),prectically insoluble in conc. h2so4 and acetone,insoluble matter in hcl: <=0.02%
Formula Weight 143.45
Physical Form Powder
Percent Purity 98%
Grade Extra Pure
Density 4.7100g/mL
Insoluble Matter 0.05% max. (in HCl)
Iron (Fe) 500ppm max.
Packaging Glass bottle
Solubility Solubility in water: soluble in cold water. Other solubilities: decomposes in hot water, sol. in hcl, hbr, nh4oh (formation of complexes), prectically insoluble in conc. h2so4 and acetone, insoluble matter in hcl: ≤0.02%
Specific Gravity 4.71
Sulfate 5000ppm max.
Boiling Point 1345.0°C
Color Green
Melting Point 504.0°C
Quantity 250g
Chemical Name or Material Copper(I) bromide
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 232-131-6

TSCA : TSCA