Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Copper(I) Bromide Dimethyl Sulfide Complex

Catalog No. 23050210G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-050-210G 10 g
23-050-250G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-050-210G Supplier Sigma Aldrich Supplier No. 23050210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Copper(I) Bromide Dimethyl Sulfide Complex

Specifications

CAS 54678-23-8
Melting Point 132°C
Molecular Formula C2H6BrCuS
Linear Formula CuBr -+ CH3SCH3
MDL Number MFCD00043295
Quantity 10 g
Synonym Copper(I) bromide methyl sulfide complex; Cuprous bromide dimethyl sulfide complex; Cuprous bromide methyl sulfide complex
Molecular Weight (g/mol) 205.58 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.