Learn More
Strem, An Ascensus Company CAS# 7787-70-4. 250g. Copper(I) bromide, min. 98%. MFCD00010969
SDP
Catalog No. 5090114861
Encompass
CAS# 7787-70-4. 250g. Copper(I) bromide, min. 98%. MFCD00010969. Molecular Weight: 143.45. Molecular Formula: CuBr. Color/form: off-white to pale-green pwdr. Strem# 93-2907. http://www.strem.com/catalog/v/93-2907/
Catalog No. 50-901-14861 Supplier Strem, An Ascensus Company Supplier No. 932907250G
CAS# 7787-70-4. 250g. Copper(I) bromide, min. 98%. MFCD00010969. Molecular Weight: 143.45. Molecular Formula: CuBr. Color/form: off-white to pale-green pwdr. Strem# 93-2907. http://www.strem.com/catalog/v/93-2907/