Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(I) bromide, Puratronic™, 99.998% (metals basis)

Catalog No. AA4075203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4075203 1 g
AA4075206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4075203 Supplier Thermo Scientific Chemicals Supplier No. 04075203
Add to Cart

CAS: 7787-70-4 | BrCu | 143.45 g/mol

It is used as effective catalyst for tetrahydropyranylation of alcohols. It can also be used in combination with palladium in catalytic synthesis of 3-haloindoles through annulation process. It is widely used in the synthesis of organic fine chemicals. The compound is widely used in the synthesis of organic compounds. It is also useful for copper-mediated coupling reactions. A silica gel support copper bromide hexa methyltriethylenetetramine complex has shown useful application for the atom transfer radical polymerization of methyl methacrylate in toluene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-70-4
Molecular Formula BrCu
Molecular Weight (g/mol) 143.45
MDL Number MFCD00010969
InChI Key NKNDPYCGAZPOFS-UHFFFAOYSA-M
Synonym copper i bromide, copper bromide cubr, copper 1+ bromide, copper monobromide, cuprousbromide, hsdb 270, cubr, copper 1 bromide
PubChem CID 24593
IUPAC Name bromocopper
SMILES [Cu+].[Br-]

Specifications

Boiling Point 1345°C
Melting Point 492°C
Color Green
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.998% (Metal basis)
Solubility Information Soluble in hydrogen bromide,hydrochloric acid,and ammonium hydroxide. Slightly soluble in water. Insoluble in acetone.
Formula Weight 143.45
Sensitivity Air Sensitive
Appearance Light green powder
Density 4.98 g/cm3
Chemical Name or Material Copper(I) bromide
Show More Show Less
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation DOT Class: 9, Packing Group: III
EINECSNumber 232-131-6
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.