Learn More
Strem, An Ascensus Company Copper(I) chloride (99.99%-Cu) PURATREM; [7758-89-6]; CuCl; Mol Wt:98.99; Color & Form: light-gray to pale green solid; Size: 50g
SDP
Catalog No. 501385470
Encompass
Copper(I) chloride (99.99%-Cu) PURATREM; [7758-89-6]; CuCl; Mol Wt:98.99; Color & Form: light-gray to pale green solid; Size: 50g
Catalog No. 50-138-5470 Supplier Strem, An Ascensus Company Supplier No. 29036050G
Copper(I) chloride (99.99%-Cu) PURATREM; [7758-89-6]; CuCl; Mol Wt:98.99; Color & Form: light-gray to pale green solid; Size: 50g