Learn More
Strem, An Ascensus Company CAS# 7758-89-6. 1kg. Copper(I) chloride, anhydrous, 97+%. MFCD00010971
SDP
Catalog No. 5090114874
Encompass
CAS# 7758-89-6. 1kg. Copper(I) chloride, anhydrous, 97+%. MFCD00010971. Molecular Weight: 98.99. Molecular Formula: CuCl. Color/form: light brown pwdr. Strem# 93-2914. http://www.strem.com/catalog/v/93-2914/
Catalog No. 50-901-14874 Supplier Strem, An Ascensus Company Supplier No. 9329141000G
CAS# 7758-89-6. 1kg. Copper(I) chloride, anhydrous, 97+%. MFCD00010971. Molecular Weight: 98.99. Molecular Formula: CuCl. Color/form: light brown pwdr. Strem# 93-2914. http://www.strem.com/catalog/v/93-2914/